missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MRI1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15827273
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15827273 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15827273 Supplier Invitrogen™ Supplier No. PA568859

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 93%; Dog: 79%; Horse: 86%; Human: 86%; Mouse: 100%; Pig: 93%; Rabbit: 79%; Rat: 86%.

MGC3207 catalyzes the interconversion of methylthioribose-1-phosphate (MTR-1-P) into methylthioribulose-1-phosphate (MTRu-1-P). Independently from catalytic activity, MGC3207 promotes cell invasion in response to constitutive RhoA activation by promoting FAK tyrosine phosphorylation and stress fiber turnover.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRI1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene MRI1
Gene Accession No. Q9CQT1
Gene Alias 2410018C20Rik; CH73-131E21.1; HAMAP-Rule:MF_03119}; M1Pi; M1Pi {ECO:0000255; mediator of RhoA-dependent invasion; Methylthioribose-1-phosphate isomerase; methylthioribose-1-phosphate isomerase {ECO:0000255; methylthioribose-1-phosphate isomerase 1; methylthioribose-1-phosphate isomerase homolog; methylthioribose-1-phosphate isomerase homolog (S. cerevisiae); MGC3207; MRDI; Mri1; MTNA; MTR-1-P isomerase; MTR-1-P isomerase {ECO:0000255; RGD1307789; similar to CG11334-PB, isoform B; S-methyl-5-thioribose-1-phosphate isomerase; S-methyl-5-thioribose-1-phosphate isomerase {ECO:0000255; S-methyl-5-thioribose-1-phosphate isomerase 1; translation initiation factor eIF-2B subunit alpha/beta/delta-like protein; translation initiation factor eIF-2B subunit alpha/beta/delta-like protein {ECO:0000255; UNQ6390/PRO21135; Ypr118w; zgc:172216
Gene Symbols MRI1
Host Species Rabbit
Immunogen synthetic peptide directed towards the N terminal region of mouse MRI1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 67873
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.