missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MRO Polyclonal Antibody
GREENER_CHOICE

Product Code. 15752481 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15752481 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15752481 Supplier Invitrogen™ Supplier No. PA554550

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: DDENDSLRYS AFVLFGQLAA FAGRKWKKFF TSQVKQTRDS LLIHLQDRNP QVAKACKTTF QACSPYLKLK EEYSFQSEED QRNTKLYQQL SHYHPEILQF FYANKIL Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 78%.

This gene is specifically transcribed in males before and after differentiation of testis, and the encoded protein may play an important role in a mammalian sex determination. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRO
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MRO
Gene Accession No. Q9BYG7
Gene Alias B29; beside the Ma29 deletion; C18orf3; maestro; male-specific transcription in the developing reproductive organs; MRO; Protein B29; Protein maestro
Gene Symbols MRO
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MRO. Recombinant protein control fragment (Product #RP-91433).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83876
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.