missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRP5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19503-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the MRP subfamily which is involved in multi-drug resistance. This protein functions in the cellular export of its substrate, cyclic nucleotides. This export contributes to the degradation of phosphodiesterases and possibly an elimination pathway for cyclic nucleotides. Studies show that this protein provides resistance to thiopurine anticancer drugs, 6-mercatopurine and thioguanine, and the anti-HIV drug 9-(2-phosphonylmethoxyethyl)adenine.Specifications
| MRP5 | |
| Polyclonal | |
| Unconjugated | |
| ABCC5 | |
| ABC33, ABCC5, MOAT C, MOATC, MRP5, pABC11, SMRP | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10057, 116721, 27416 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.46 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O15440, Q9QYM0, Q9R1X5 | |
| Abcc5 | |
| Peptide | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction