Learn More
MRPL24 Mouse, anti-Human, Clone: 3D3C8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68556-1-IG-150UL
This item is not returnable.
View return policy
Description
The MRPL24 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human MRPL24. This antibody recognizes human antigen. The MRPL24 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| MRPL24 | |
| 3D3C8 | |
| FLJ20917, L24mt, MRP L18, MRP L24, MRPL24 | |
| 150 μL | |
| 79590 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, ELISA | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG1 |