missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MRPL41 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15743031 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15743031 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15743031 Supplier Invitrogen™ Supplier No. PA555105

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: KPYVSYLAPE SEETPLTAAQ LFSEAVAPAI EKDFKDGTFD PDNLEKYGFE PTQEGKLFQL YPRNFLR Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 78%.

Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein that belongs to the YmL27 ribosomal protein family. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRPL41
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MRPL41
Gene Accession No. Q8IXM3
Gene Alias 39S ribosomal protein L27 homolog; 39S ribosomal protein L41, mitochondrial; Bcl-2-interacting mitochondrial ribosomal protein L41; BMRP; Cell proliferation-inducing gene 3 protein; L41mt; Mitochondrial large ribosomal subunit protein mL41; mitochondrial ribosomal protein L41; MRPL27; MRP-L27; MRP-L27 homolog; Mrpl41; MRP-L41; PIG3; proliferation-inducing gene 3; Rpml27
Gene Symbols MRPL41
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MRPL41. Recombinant protein control fragment (Product #RP-93110).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64975
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.