missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPS15 Antibody (OTI6D2), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
256.00 € - 550.00 €
Specifications
| Antigen | MRPS15 |
|---|---|
| Clone | OTI6D2 |
| Dilution | Western Blot 1:2000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30306219
|
Novus Biologicals
NBP3-44909-30ul |
30 μL |
256.00 €
30µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30306320
|
Novus Biologicals
NBP3-44909-100ul |
100 μL |
550.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MRPS15 Monoclonal antibody specifically detects MRPS15 in Human samples. It is validated for Western BlotSpecifications
| MRPS15 | |
| Western Blot 1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3) 1% BSA, 50% glycerol | |
| 64960 | |
| IgG2b | |
| >95% | |
| Antibody |
| OTI6D2 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| FLJ11564,28S ribosomal protein S15, mitochondrial, mitochondrial ribosomal protein S15, MPR-S15, MRP-S15, RPMS15, S15mt | |
| Full length human recombinant protein of human MRPS15(NP_112570) produced in E.coli. | |
| Primary | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title