Learn More
Invitrogen™ MRPS15 Recombinant Rabbit Monoclonal Antibody (24GB70)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557481
 View more versions of this product
Product Code. 30276494
  • 198.00 € / 20µL
Description
MRPS15 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. MRPS15 encodes a 28S subunit protein that belongs to the ribosomal protein S15P family. The encoded protein is more than two times the size of its E. coli counterpart, with the 12S rRNA binding sites conserved. Between human and mouse, the encoded protein is the least conserved among small subunit ribosomal proteins. Pseudogenes corresponding to this gene are found on chromosomes 15q and 19q.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MRPS15
Recombinant Monoclonal
Unconjugated
MRPS15
1500003E24Rik; 2410002B11Rik; 28S ribosomal protein S15, mitochondrial; DC37; FLJ11564; mitochondrial ribosomal protein S15; Mitochondrial small ribosomal subunit protein uS15m; Mprs15; MPR-S15; Mrps15; MRP-S15; RPMS15; S15mt
Rabbit
Affinity chromatography
RUO
64960
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB70
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P82914
MRPS15
A synthesized peptide derived from human MRPS15 (aa 200-257).
20 μL