missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPS22 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
288.00 € - 528.00 €
Specifications
| Antigen | MRPS22 |
|---|---|
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Regulatory Status | RUO |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18435631
|
Novus Biologicals
NBP1-86787-25ul |
25 μL |
288.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18210866
|
Novus Biologicals
NBP1-86787 |
0.1 mL |
528.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MRPS22 Polyclonal specifically detects MRPS22 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.Specifications
| MRPS22 | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 56945 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:TKKPTFMDEEVQSILTKMTGLNLQKTFKPAIQELKPPTYKLMTQAQLEEATRQAVEAAKVRLKMPPVLEERVPINDVLAEDKILEGTETTKYVFTDISYSIPHRERFIVVREPSGTLRKA | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Polyclonal | |
| Rabbit | |
| Human, Mouse, Rat | |
| C3orf5GK002, COXPD5, GIBT, mitochondrial ribosomal protein S22, MRP-S2228S ribosomal protein S22, mitochondrial, RPMS22, S22mt | |
| MRPS22 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title