missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPS28 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93219-0.1ml
This item is not returnable.
View return policy
Description
MRPS28 Polyclonal antibody specifically detects MRPS28 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| MRPS28 | |
| Polyclonal | |
| Western Blot 1:500-1:1000, Immunocytochemistry/ Immunofluorescence 1:20-1:100 | |
| 28S ribosomal protein S35, mitochondrial, FLJ22853, mitochondrial 28S ribosomal protein S35, mitochondrial ribosomal protein S28, MRP-S2828S ribosomal protein S28, mitochondrial, MRP-S35, MRPS35HSPC007, S28mt, S35mt | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-187 of human MRPS28 (NP_054737.1). MAALCRTRAVAAESHFLRVFLFFRPFRGVGTESGSESGSSNAKEPKTRAGGFASALERHSELLQKVEPLQKGSPKNVESFASMLRHSPLTQMGPAKDKLVIGRIFHIVENDLYIDFGGKFHCVCRRPEVDGEKYQKGTRVRLRLLDLELTSRFLGATTDTTVLEANAVLLGIQESKDSRSKEEHHEK | |
| 0.1 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 28957 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction