missing translation for 'onlineSavingsMsg'
Learn More

MRRF Monoclonal Antibody (OTI5C11), TrueMAB™, OriGene

Product Code. 17556443
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17556443 100 μL 100µL
17576443 30 μL 30µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17556443 Supplier Origene Technologies Supplier No. TA806313

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MRRF is responsible for the release of ribosomes from messenger RNA at the termination of protein biosynthesis, may increase the efficiency of translation by recycling ribosomes from one round of translation to another.

Specifications

Antigen MRRF
Applications Western Blot
Classification Monoclonal
Clone OTI5C11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene MRRF
Gene Accession No. Q96E11
Gene Alias MRFF, MTRRF, RRF
Gene Symbols MRRF
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 56-262 of human MRRF produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 92399
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.