missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: EALKDNFNKT LNIRTSPGSL DKIAVVTADG KLALNQISQI SMKSPQLILV NMASFPECTA AAIKAIRESG MNLNPEVEGT LI Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 95%.
Responsible for the release of ribosomes from messenger RNA at the termination of protein biosynthesis. May increase the efficiency of translation by recycling ribosomes from one round of translation to another. Ota T., Nat. Genet. 36:40-45(2004). Humphray S.J., Nature 429:369-374(2004). Zhang Y., Biochim. Biophys. Acta 1443:245-250(1998).
Specifications
Specifications
| Antigen | MRRF |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | Mrrf |
| Gene Accession No. | Q96E11 |
| Gene Alias | 2400002D02Rik; mitochondrial ribosome recycling factor; MRFF; MRRF; MTRRF; Ribosome-recycling factor, mitochondrial; ribosome-releasing factor, mitochondrial; RRF |
| Gene Symbols | Mrrf |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
