missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MSH3 Antibody (CL14059) [PE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-24587PE
This item is not returnable.
View return policy
Description
MSH3 Monoclonal antibody specifically detects MSH3 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| MSH3 | |
| Monoclonal | |
| PE | |
| Divergent upstream protein, DNA mismatch repair protein Msh3, DUC1, DUG, DUPMGC163306, hMSH3, Mismatch repair protein 1, MRP1MGC163308, mutS (E. coli) homolog 3, mutS homolog 3 (E. coli) | |
| This antibody was developed using a synthetic peptide derived from P20585, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| DNA Repair, Mismatch Repair, Neuroscience | |
| 4437 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CL14059 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction