Learn More
MSH3 Antibody (CL14064) [PerCP], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-24588PCP
Product Code. 30300739
  • 737.00 € / 0.10mL
Description
MSH3 Monoclonal antibody specifically detects MSH3 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MSH3
Monoclonal
PerCP
Divergent upstream protein, DNA mismatch repair protein Msh3, DUC1, DUG, DUPMGC163306, hMSH3, Mismatch repair protein 1, MRP1MGC163308, mutS (E. coli) homolog 3, mutS homolog 3 (E. coli)
This antibody was developed using a synthetic peptide derived from P20585, with the exact immunogen sequence remaining proprietary.
0.1 mL
DNA Repair, Mismatch Repair, Neuroscience
4437
Store at 4⪚C in the dark.
IgG2b
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
CL14064
PBS
Mouse
Protein A purified
RUO
Primary
Human
Purified