missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MSH6 Antibody (MSH6/7064R) - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20784
This item is not returnable.
View return policy
Description
MSH6 Monoclonal antibody specifically detects MSH6 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)
Specifications
| MSH6 | |
| Monoclonal | |
| 1 mg/mL | |
| ELISA, Immunohistochemistry-Paraffin 1-2 μg/mL | |
| DNA mismatch repair protein Msh6, G/T mismatch-binding protein, GTBPHNPCC5, GTMBP, hMSH6, HSAP, mutS (E. coli) homolog 6, mutS homolog 6 (E. coli), MutS-alpha 160 kDa subunit, p160, sperm-associated protein | |
| Recombinant fragment (around aa 374-540) of human MSH6 protein (exact sequence is proprietary) | |
| 100 μg | |
| Breast Cancer, Cancer, DNA Repair, Mismatch Repair | |
| 2956 | |
| Store at -20°C to -70°C. Avoid freeze/thaw cycles. | |
| IgG κ |
| ELISA, Immunohistochemistry (Paraffin) | |
| MSH6/7064R | |
| Unconjugated | |
| 10 mM PBS | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction