Learn More
Invitrogen™ MSP Recombinant Rabbit Monoclonal Antibody (24GB635)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557054
 View more versions of this product
Product Code. 30276509
  • 146.00 € / 20µL
Description
MSP Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
MSP (or MST1) contains four kringle domains and a serine protease domain, similar to that found in hepatic growth factor. Despite the presence of the serine protease domain, the encoded protein may not have any proteolytic activity. The receptor for this protein is RON tyrosine kinase, which upon activation stimulates ciliary motility of ciliated epithelial lung cells. This protein is secreted and cleaved to form an alpha chain and a beta chain bridged by disulfide bonds.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MSP
Recombinant Monoclonal
Unconjugated
MST1
D3F15S2; D3F15S2h; D8h3f15s2; D9H3F15S2; DNF15S2; DNF15S2h; E2F transcription factor 2; E2F2; Hepatocyte growth factor-like protein; Hepatocyte growth factor-like protein alpha chain; Hepatocyte growth factor-like protein beta chain; hepatocyte growth factor-like protein homolog; Hgfl; macrophage stimulating 1; macrophage stimulating 1 (hepatocyte growth factor-like); macrophage stimulatory protein; Macrophage-stimulating protein; MSP; Mst1; NF15S2; Unknown (protein for MGC:137619)
Rabbit
Affinity chromatography
RUO
15235, 24566, 4485
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB635
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P26927, P26928
MST1
A synthesized peptide derived from human MST1 (aa 15-65).
20 μL
Primary
Human, Mouse, Rat