Learn More
MST1/STK4 Antibody (07), Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-06389-50ul
 View more versions of this product
Product Code. 18679788
  • 220.00 € / 50µL
Description
MST1/STK4 Monoclonal antibody specifically detects MST1/STK4 in Human samples. It is validated for ELISA, Sandwich ELISA Capture
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MST1/STK4
Monoclonal
Unconjugated
AI447339;AU020804;Kas-2;KRS2;MST1;STK4;YSK3
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human MST1/STK4 (Uniprot#: Q13043-1; Glu2-Phe487).
50 μg
Apoptosis, Autophagy, Cardiovascular Biology, Cell Biology, Signal Transduction
Human
Purified
ELISA, Sandwich ELISA Capture
07
0.2 um filtered solution in PBS
Mouse
Protein A purified
RUO
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1