missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MSX2 Mouse, anti-Human, Clone: 1B1G3, Proteintech
Mouse Monoclonal Antibody
145.00 € - 410.00 €
Specifications
| Antigen | MSX2 |
|---|---|
| Clone | 1B1G3 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, ELISA |
| Form | Liquid |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18078175
|
Proteintech
68550-1-IG-150UL |
150 μL |
410.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18046395
|
Proteintech
68550-1-IG-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The MSX2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human MSX2. This antibody recognizes human antigen. The MSX2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| MSX2 | |
| 1000 μg/mL | |
| Liquid | |
| Human | |
| CRS2, FPP, Homeobox protein Hox 8, Homeobox protein MSX 2, HOX8, MSH, msh homeobox 2, MSX2, PFM, PFM1 | |
| Primary |
| 1B1G3 | |
| Western Blot, ELISA | |
| Mouse | |
| 4488 | |
| IgG1 | |
| Store at -20°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title