missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MT-ND5 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-05606-100ul
This item is not returnable.
View return policy
Description
MT-ND5 Polyclonal antibody specifically detects MT-ND5 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| MT-ND5 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin | |
| EC 1.6.5.3, mitochondrially encoded NADH dehydrogenase 5, MTND5, NAD5, NADH dehydrogenase subunit 5, NADH dehydrogenase, subunit 5 (complex I), NADH5, ND5NADH dehydrogenase 5 | |
| A synthetic peptide corresponding to a sequence within amino acids 500-607 of mouse MT-ND5 (NP_904338.1). IALELNNLTMKLSMNKANPYSSFSTLLGFFPSIIHRITPMKSLNLSLKTSLTLLDLIWLEKTIPKSTSTLHTNMTTLTTNQKGLIKLYFMSFLINIILIIILYSINLE | |
| 100 μg | |
| Cancer, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction | |
| 4540 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction