missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTAP Antibody (MTAP/1813), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-75730-100ug
This item is not returnable.
View return policy
Description
MTAP Monoclonal antibody specifically detects MTAP in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array
Specifications
| MTAP | |
| Monoclonal | |
| 0.2 mg/mL | |
| Western Blot 1-2 μg/mL, ELISA 2-4 μg/mL, Immunohistochemistry, Immunohistochemistry-Paraffin 1-2 μg/mL, Protein Array | |
| Q13126 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 4507 | |
| Store at 4°C. | |
| IgG2b κ |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array | |
| MTAP/1813 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| 5'-methylthioadenosine phosphorylase, c86fus, EC 2.4.2.28, methylthioadenosine phosphorylase, MSAPMeSAdo phosphorylase, MTA phosphorylase, MTAPase, S-methyl-5'-thioadenosine phosphorylase | |
| Recombinant human MTAP protein fragment (aa97-196) (exact sequence is proprietary) (Uniprot: Q13126) | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction