missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTAP (Tumor Suppressor Marker) Mouse, Clone: MTAP/1813, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 4507-MSM1-P1
This item is not returnable.
View return policy
Specifications
| MTAP (Tumor Suppressor Marker) | |
| Cytoplasmic | |
| MTAP/1813 | |
| Unconjugated | |
| Mouse | |
| 31kDa | |
| Protein A/G purified from Bioreactor concentrate. | |
| RUO | |
| 4507 | |
| IgG2b κ |
| ELISA, Western Blot, Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
| Recombinant human MTAP protein fragment (aa97-196) | |
| HepG2, A431, HeLa or MCF-7 cells. Kidney | |
| 100 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction