Learn More
MTCO2 Mouse, anti-Human, Clone: 10G14F4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68527-1-IG-20UL
This item is not returnable.
View return policy
Description
The MTCO2 antibody from Proteintech is a mouse monoclonal antibody generated with peptide of human MTCO2. This antibody recognizes human antigen. The MTCO2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| MTCO2 | |
| 10G14F4 | |
| COII, COX2, COXII, cytochrome c oxidase II, Cytochrome c oxidase subunit 2, MT CO2, MTCO2 | |
| 20 μL | |
| 4513 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, ELISA | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG1 |