missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTERFD3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94738-0.02ml
This item is not returnable.
View return policy
Description
MTERFD3 Polyclonal antibody specifically detects MTERFD3 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| MTERFD3 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000 | |
| FLJ14062, Mitochondrial transcription termination factor 2, Mitochondrial transcription termination factor-like protein, MTERF domain containing 3, mTERF domain-containing protein 3, mitochondrial, mTERF2, mTERFL | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-250 of human MTERFD3 (NP_079474.2). MLWKLLLRSQSCRLCSFRKMRSPPKYRPFLACFTYTTDKQSSKENTRTVEKLYKCSVDIRKIRRLKGWVLLEDETYVEEIANILQELGADETAVASILERCPEAIVCSPTAVNTQRKLWQLVCKNEEELIKLIEQFPESFFTIKDQENQKLNVQFFQELGLKNVVISRLLTAAPNVFHNPVEKNKQMVRILQESYLDVGGSEANMKVWLLKLLSQNPFILLNSPTAIKETLEFLQEQGFTSFEILQLLSK | |
| 0.02 mL | |
| Endocrinology | |
| 80298 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction