missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTRF1L Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-85893
This item is not returnable.
View return policy
Description
MTRF1L Polyclonal antibody specifically detects MTRF1L in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| MTRF1L | |
| Polyclonal | |
| Western Blot 0.04-0.4 ug/mL, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| mitochondrial translational release factor 1-likemitochondrial, MRF1L, MTRF1A | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: KELAMTKLRAKLYSMHLEEEINKRQNARKIQIGSKGRSEKIRTYNFPQNRVTDHRINKTLHDLETFMQGDYLLDELVQSLKE | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 54516 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction