missing translation for 'onlineSavingsMsg'
Learn More
Learn More
mtTFA Antibody (TFAM/9713), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-44621-100ug
This item is not returnable.
View return policy
Description
mtTFA Monoclonal antibody specifically detects mtTFA in Human samples. It is validated for Immunohistochemistry,Western Blot
Specifications
| mtTFA | |
| Monoclonal | |
| 0.2 mg/ml | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Western Blot | |
| TFAM/9713 | |
| Unconjugated | |
| mitochondrial transcription factor A, MtTF1, mtTFA, TCF-6, TCF6L1, TCF6L2Mitochondrial transcription factor 1, TCF6TCF6L3, Transcription factor 6, Transcription factor 6-like 2, Transcription factor 6-like 2 (mitochondrial transcription factor), transcription factor A, mitochondrial | |
| Recombinant fragment (around aa1-200) of human mtTFA protein (Uniprot: Q00059). Exact sequence is proprietary. | |
| 100 μg | |
| Cell Cycle and Replication, Transcription Factors and Regulators | |
| 7019 | |
| Store at 4⪚C. | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction