missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTTP Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
216.00 € - 493.00 €
Specifications
| Antigen | MTTP |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18608012
|
Novus Biologicals
NBP2-94915-0.02ml |
0.02 mL |
216.00 €
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18613252
|
Novus Biologicals
NBP2-94915-0.1ml |
0.1 mL |
493.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MTTP Polyclonal antibody specifically detects MTTP in Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
| MTTP | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cardiovascular Biology, Endocrinology, Signal Transduction | |
| PBS (pH 7.3), 50% glycerol | |
| 4547 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse, Rat | |
| 88kD), 88kDa), ABL, MGC149819, microsomal triglyceride transfer protein, microsomal triglyceride transfer protein large subunit, MTPMGC149820 | |
| A synthetic peptide corresponding to a sequence within amino acids 550-650 of human MTTP (NP_000244.2). SKIVRRVLKEMVAHNYDRF | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title