missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTUS2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-86604
This item is not returnable.
View return policy
Description
MTUS2 Polyclonal specifically detects MTUS2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| MTUS2 | |
| Polyclonal | |
| Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:20 - 1:50, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| Cardiac zipper protein, CAZIPTracking protein of 150 kDa, KIAA0774+TIP of 150 kDa, microtubule associated tumor suppressor candidate 2, Microtubule plus-end tracking protein TIP150, TIP150microtubule-associated tumor suppressor candidate 2 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 23281 | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| MTUS2 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:DSVLNIPAPLHPETTVNMTYQPTTPSSSFQDVSVFGMDAGSPLVVPPPTDSARLLNTSPKVPDKNTCPSGIPKPVFTHSKDTPSSQEGMENYQV | |
| 0.1 mL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction