missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MUC13 Antibody (MUC13/13112), Novus Biologicals™
Mouse Monoclonal Antibody
256.00 € - 484.00 €
Specifications
| Antigen | MUC13 |
|---|---|
| Clone | MUC13/13112 |
| Concentration | 0.2 mg/ml |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30286715
|
Novus Biologicals
NBP3-44616-20ug |
20 μg |
256.00 €
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30290524
|
Novus Biologicals
NBP3-44616-100ug |
100 μg |
484.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MUC13 Monoclonal antibody specifically detects MUC13 in Human samples. It is validated for ImmunohistochemistrySpecifications
| MUC13 | |
| 0.2 mg/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| 56667 | |
| IgG1 κ | |
| Protein G purified |
| MUC13/13112 | |
| Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| Down-regulated in colon cancer 1FLJ20063, DRCC1, MUC-13, mucin 13, cell surface associated, mucin 13, epithelial transmembrane, mucin-13, RECC | |
| Recombinant fragment (around aa100-300) of human MUC13 protein (Uniprot: Q9H3R2). Exact sequence is proprietary. | |
| Primary | |
| Store at 4⪚C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title