Learn More
Invitrogen™ MUC17 Biosimilar Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA559350
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °. This antibody can also be cross referenced by the term Forerunner patent anti-Muc 17.
MUC17, a member of the mucin family of glycosylated proteins, is located in the q22.1 region of chromosome 7, specifically between the MUC12 and SERPINE1 genes. The MUC17 gene spans a 39-kb DNA fragment and contains 13 exons, producing a 14.2-kb mRNA. MUC17 is characterized by the generation of two splice variants that encode either a membrane-anchored or a secreted form. This gene plays a crucial role in epithelial cell protection and renewal, primarily in the gastrointestinal tract. MUC17's large extracellular domain, which includes tandem repeat regions, is extensively glycosylated, contributing to the formation of a protective mucous barrier.
Specifications
| MUC17 Biosimilar | |
| Recombinant Monoclonal | |
| Unconjugated | |
| Human | |
| 1 mg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
| 1 mg/mL | |
| 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
| Protein A | |
| RUO | |
| Human | |
| Antibody | |
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.