missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MUC17 Recombinant Human Monoclonal Antibody (8D12)

Description
MUC17 Recombinant Monoclonal Antibody for ELISA
Specifications
Specifications
| Antigen | MUC17 |
| Applications | ELISA |
| Classification | Recombinant Monoclonal |
| Clone | 8D12 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 |
| Gene | MUC17 |
| Gene Accession No. | Q685J3 |
| Gene Alias | membrane mucin MUC17; MUC17; MUC-17; MUC3; MUC-3; mucin 17, cell surface associated; mucin-17; secreted mucin MUC17; small intestinal mucin MUC3; small intestinal mucin-3 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?