Learn More
MUC2 Antibody (CL9148) [Alexa Fluor™ 532], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-21168AF532
Product Code. 30307913
  • 737.00 € / 0.10mL
Description
MUC2 Monoclonal antibody specifically detects MUC2 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MUC2
Monoclonal
Alexa Fluor 532
Intestinal mucin-2, MLP, MUC2, MUC-2, mucin 2, intestinal/tracheal, mucin 2, oligomeric mucus/gel-forming, mucin-2, SMUC
This antibody was developed using a synthetic peptide derived from Q02817, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cancer, Cellular Markers, Inflammation, Signal Transduction
4583
Store at 4⪚C in the dark.
IgG2a
Immunohistochemistry, Immunohistochemistry (Paraffin)
CL9148
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified