missing translation for 'onlineSavingsMsg'
Learn More

MUC5AC Antibody (rMUC5AC/8052), Novus Biologicals™

Mouse Monoclonal Antibody

232.00 € - 462.00 €

Specifications

Antigen MUC5AC
Clone rMUC5AC/8052
Concentration 0.2 mg/mL
Dilution Immunohistochemistry-Paraffin : 1-2 μg/mL
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18687754
View Documents
Novus Biologicals
NBP3-23947-20ug
20 μg
232.00 €
20µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18653665
View Documents
Novus Biologicals
NBP3-23947-100ug
100 μg
462.00 €
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

MUC5AC Monoclonal antibody specifically detects MUC5AC in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

MUC5AC
0.2 mg/mL
Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Cellular Markers, Extracellular Matrix, Infections (Virus Bacteria and Parasites), Signal Transduction
10 mM PBS with 0.05% BSA
4586
IgG1 κ
Protein A or G purified
rMUC5AC/8052
Immunohistochemistry-Paraffin : 1-2 μg/mL
Monoclonal
Purified
RUO
Human
gastric mucin, leB, lewis B blood group antigen, major airway glycoprotein, MUC5, mucin 5, subtypes A and C, tracheobronchial/gastric, mucin 5AC, oligomeric mucus/gel-forming, mucin 5AC, oligomeric mucus/gel-forming pseudogene, mucin-5 subtype AC, tracheobronchial, mucin-5AC, TBM, tracheobronchial mucin
Recombinant full-length human MUC5AC protein
Primary
Store at 4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.