missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MUC5AC Monoclonal Antibody (2-12M1)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA135705

Product Code. 11525012

  • 520.00 € / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

2-12M1 recognizes the peptide core of gastric mucin M1/MUC5AC, and more specifically with the c epitope amongst the a, b, c, d, e, f, g, and h protein core epitopes defined by Bara for M1. 2-12M1 and 45M1 both specifically react with epitopes located in the Cterminal cysteine rich part of the peptide core of gastric mucin (MUC5AC). MUC5AC is present in primary ovarian mucinous cancer and gastric cancer, but usually absent in colorectal adenocarcinoma, thus showing an expression pattern opposite to MUC2. AntiMUC5AC may be useful for differential identification of primary mucinous ovarian tumors from colon adenocarcinoma metastatic to the ovary. MUC5AC antibodies may also be useful for identification pancreatic carcinoma and pre-cancerous changes vs. normal pancreas.

The antibody 2-25LE reacts with Lewisb blood group antigen, a carbohydrate determinant carried on both glycolipids and glycoproteins, detected on erythrocytes, certain epithelial cells, and in secretions of certain individuals.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MUC5AC
Monoclonal
0.1 mg/mL
PBS with 0.02% sodium azide
P98088
MUC5AC
Mucin isolated from ovarian cyst fluid.
100 μg
Primary
Human, Mouse, Monkey, Bovine, Feline
Antibody
IgG1
ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
2-12M1
Unconjugated
MUC5AC
2210005L13Rik; gastric mucin; LeB; lewis B blood group antigen; LOW QUALITY PROTEIN: mucin-5AC; major airway glycoprotein; MGM; MUC5; MUC5AC; MUC-5AC; mucin; mucin 5, subtypes A and C; mucin 5, subtypes A and C, tracheobronchial/gastric; mucin 5ac; mucin 5AC, oligomeric mucus/gel-forming; mucin 5AC, oligomeric mucus/gel-forming pseudogene; mucin-5 subtype AC, tracheobronchial; Mucin-5AC; TBM; Tracheobronchial mucin
Mouse
Purified
RUO
100847395, 105261014, 17833, 4586
4°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.