missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mucin 19 Monoclonal Antibody (876013)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561931
This item is not returnable.
View return policy
Description
Mucin 19 Monoclonal Antibody for IHC (P)
This gene encodes a member of the gel-forming mucin protein family. Mucin family members are glycoproteins that have tandem repeats which are extensively O-glycosylated. The structural features of mucin proteins are responsible for the gel-like properties of mucus. The encoded protein may be involved in disruption of the ocular surface in Sjogren syndrome. [provided by RefSeq, Apr 2014].
Specifications
| Mucin 19 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with no preservative | |
| Q7Z5P9 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 283463 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Immunohistochemistry (Paraffin) | |
| 876013 | |
| Unconjugated | |
| MUC19 | |
| MUC19 | |
| E. coli-derived recombinant human MUC-19, Lys5606-Gly5767. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction