missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Mueller-Hinton Agar with 5% Sheep Blood

Product Code. 12917148
Change view
Click to view available options
Fill Weight:
26.0g ±1.0.g
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Fill Weight unitSize
12917148 26.0g ±1.0.g Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12917148 Supplier Thermo Scientific™ Supplier No. PB0431A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Perform effortless Antimicrobial Susceptibility Tests (AST) for fastidious organisms using prepared plates which conform to CLSI standard.

Perform effortless Antimicrobial Susceptibility Tests (AST) for fastidious organisms using Thermo Scientific™ Mueller-Hinton Agar with 5% Sheep Blood prepared media plates. A precise clear zone will help to determine if the bacteria is resistant or sensitive to the antibiotics used. Mueller-Hinton Agar with 5% Sheep Blood conforms to the American standard of the Clinical and Laboratory Standards Institute (CLSI) and European Standard of Deutsches Institut für Normung (DIN).

The major use of Mueller-Hinton Agar with 5% Sheep Blood is for Antimicrobial Susceptibility Testing (AST). The medium is designed and manufactured according to CLSI standards and can be used in internationally recognised standard procedures.

The inclusion of starch in the medium ensures that toxic factors produced during microbial growth will be absorbed, and its presence is often essential to establish growth from very small inocula.

Not all products are available for sale in all territories. Please inquire.
TRUSTED_SUSTAINABILITY

Specifications

Color Traffic red, opaque
Content And Storage 2°C to 10°C
Description Mueller-Hinton Agar with 5% Sheep Blood
Format 90 mm Monoplate
Product Type Agar Plate
Fill Weight 26.0g ±1.0.g
Packaging Type Wrapped in single cellulose-based film wrap
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.