missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MUL1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94291-0.1ml
This item is not returnable.
View return policy
Description
MUL1 Polyclonal antibody specifically detects MUL1 in Human, Mouse samples. It is validated for Western Blot
Specifications
| MUL1 | |
| Polyclonal | |
| Western Blot 1:1000 - 1:2000 | |
| C1orf166mitochondrial E3 ubiquitin ligase 1, E3 ubiquitin-protein ligase MUL1, EC 6.3.2, EC 6.3.2.-, FLJ12875, GIDERP11-401M16.2, Growth inhibition and death E3 ligase, MAPLE3 ubiquitin ligase, mitochondrial E3 ubiquitin protein ligase 1, mitochondrial ubiquitin ligase activator of NF-kB, mitochondrial ubiquitin ligase activator of NFKB 1, Mitochondrial-anchored protein ligase, MULANchromosome 1 open reading frame 166, Putative NF-kappa-B-activating protein 266, RING finger protein 218, RNF218mitochondria-anchored protein ligase | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-352 of human MUL1 (NP_078820.2). MESGGRPSLCQFILLGTTSVVTAALYSVYRQKARVSQELKGAKKVHLGEDLKSILSEAPGKCVPYAVIEGAVRSVKETLNSQFVENCKGVIQRLTLQEHKMVWNRTTHLWNDCSKIIHQRTNTVPFDLVPHEDGVDVAVRVLKPLDSVDLGLETVYEKFHPSIQSFTDVIGHYISGERPKGIQETEEMLKVGATLTGVGELVLDNNSVRLQPPKQGMQYYLSSQDFDSLLQRQESSVRLWKVLALVFGFATCATLFFILRKQYLQRQERLRLKQMQEEFQEHEAQLLSRAKPEDRESLKSACVVCLSSFKSCVFLECGHVCSCTECYRALPEPKKCPICRQAITRVIPLYNS | |
| 0.1 mL | |
| Apoptosis, Signal Transduction, Ubiquitin Proteasome Pathway | |
| 79594 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction