missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Muller-Kauffmann Tetrathionate-Novobiocin (MkTTn) Enrichment Broth

Product Code. 12755640
Change view
Click to view available options
Fill Weight:
10 ± 1mL
Manufacturing Origin:
Germany
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Fill Weight Manufacturing Origin Unit Size
12755640 10 ± 1mL Germany Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12755640 Supplier Thermo Scientific™ Supplier No. TV5065E
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Selectively enrich Salmonella species from food with the convienience of Muller-Kauffmann Tetrathionate-Novobiocin Enrichment Broth prepared media tubes.

Selectively isolate Salmonella species with the convenience of Thermo Scientific™ Muller-Kauffmann Tetrathionate-Novobiocin (MkTTn) Enrichment Broth prepared media tubes. MkTTn is a selective enrichment medium for the isolation of Salmonella, whose formulation conforms to ISO 6579:20021 for the microbiological testing food and animal feeding stuffs.

Muller-Kauffmann Tetrathionate-Novobiocin Broth was developed by Muller2 and later modified by Kauffmann3,4 with the addition of ox bile and brilliant green to improve selectivity. The addition of novobiocin at 40 mg per liter was later described by Jeffries5 to improve inhibition of Proteus species.

After the sample has been incubated in Buffered Peptone Water (ISO), transfer 1 ml of the broth to 10 ml of MkTTn Broth and 0.1 ml into 10 ml of RVS Broth (TV5036E). Incubate the MkTTn Broth at 37°C ± 1°C for 24 hours ± 3 hours and the RVS Broth at 41.5°C ± 1°C, for 24 hours ± 3 hours. Sub-culture the incubated MkTTn and RVS broths onto XLD Agar (PO5057A/PO0164A) and a second agar medium of choice, and incubate for a further 24 hours ± 3 hours at 37°C ± 1°C. Presumptive Salmonella should be confirmed using appropriate biochemical and serological techniques. For the complete method, please refer to ISO 6579:20021.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

TRUSTED_SUSTAINABILITY

Specifications

Color Light green, opaque
Content And Storage 2°C to 12°C
Description Muller Kauffmann Tetrathionate Novobiocin Enrichment Broth
Format Tube
Manufacturing Origin Germany
Product Type Microbiology Media
Fill Volume 10 mL
Fill Weight 10 ± 1mL
Packaging Type 50 tubes in a carton box
Quantity 50/Pk.

For laboratory use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.