missing translation for 'onlineSavingsMsg'
Learn More

Multi 7 ions, 10mg/L of each Component, for Ion Chromatography, H2O matrix, Fisher chemical™
BIOPHARMA

Product Code. 12942235 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12942235 500 mL 500mL
12932235 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 12942235 Proveedor Fisher Chemical N.º de proveedor A 101 500
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Ions (7 components): F-,Cl-,NO2-,Br-,NO23-,PO43-,SO42

Specifications

Chemical Name or Material Multi 7 ions
Packaging Bottle
Analyte or Component Names F-,Cl-,NO2-,Br-,NO23-,PO43-,SO42
Physical Form Liquid
Concentration 10 mg/L of each Component
Type Multi-component Standard Solution
Element F-, Cl-, NO2-, Br-, NO23-, PO43-, SO42
Solvent or Matrix Water
Quantity 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.