missing translation for 'onlineSavingsMsg'
Learn More

Multi-Anion Standard 7A: F-, Cl-, NO3- as N, Br-, SO4(-2), PO4(-3) as P @ 1000 μg/mL in H2O, VHG™

Product Code. 17918404
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17918404 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17918404 Supplier VHG Labs Supplier No. VHGICM7A100
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Multi-Anion Standard 7A: F-, Cl-, NO3- as N, Br-, SO4(-2), PO4(-3) as P @ 1000 μg/mL in H2O
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bromide/Chloride/Fluoride/Nitrate/Phosphate/Sulfate
CAS 24959-67-9, 16887-00-6, 16984-48-8, 14797-55-8, 14265-44-2, 14808-79-8
Methods Ion Chromatography
Analyte or Component Names Bromide/Chloride/Fluoride/Nitrate/Phosphate/Sulfate
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Water
Quantity 100 mL
Concentration or Composition (by Analyte or Components) Bromide 1000 μg/mL/Chloride 1000 μg/mL/Fluoride 1000 μg/mL/Nitrate 1000 μg/mL/Phosphate 1000 μg/mL/Sulfate 1000 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.