Learn More
Multi-Element Organometallic Oil Standards 23 Element, 100μg/g; SPEX CertiPrep™
Shop All SPEX Certiprep ProductsDescription
Includes
NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
| Analyte | Concentration |
|---|---|
| Ag | 100μg/g |
| Al | 100μg/g |
| B | 100μg/g |
| Ba | 100μg/g |
| Ca | 100μg/g |
| Cd | 100μg/g |
| Cr | 100μg/g |
| Cu | 100μg/g |
| Fe | 100μg/g |
| K | 100μg/g |
| Mg | 100μg/g |
| Mn | 100μg/g |
| Mo | 100μg/g |
| Na | 100μg/g |
| Ni | 100μg/g |
| P | 100μg/g |
| Pb | 100μg/g |
| Sb | 100μg/g |
| Si | 100μg/g |
| Sn | 100μg/g |
| Ti | 100μg/g |
| V | 100μg/g |
| Zn | 100μg/g |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Spécification
Spécification
| Chemical Name or Material | 23 Element Oil Standard |
| Packaging | Triple-leached LDPE bottle |
| Concentration or Composition Notes | Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. |
| Analyte or Component Names | Al,Ba,B |
| For Use With (Equipment) | AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation |
| Type | Certified Reference Material |
| Element | Al, Ba, B |
| Solvent or Matrix | Base Oil 75 |
| Industry Type | Petrochemical Testing |
| Quantity | 50 g |
Product intended for laboratory use only
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.