missing translation for 'onlineSavingsMsg'
Learn More

Multi-Element Organometallic Oil Standards 23 Element, 900μg/g; SPEX CertiPrep™

Product Code. 11460359 Shop All SPEX Certiprep Products
Change view
Click to view available options
Quantity:
50 g
100 g
Packaging:
Triple-leached LDPE bottle
Unit Size:
100g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11460359 50 g Triple-leached LDPE bottle 50g
15246386 100 g Triple-leached LDPE bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11460359 Supplier SPEX Certiprep Supplier No. S23900Z

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Ag900μg/g
Al900μg/g
B900μg/g
Ba900μg/g
Ca900μg/g
Cd900μg/g
Cr900μg/g
Cu900μg/g
Fe900μg/g
K900μg/g
Mg900μg/g
Mn900μg/g
Mo900μg/g
Na900μg/g
Ni900μg/g
P900μg/g
Pb900μg/g
Sb900μg/g
Si900μg/g
Sn900μg/g
Ti900μg/g
V900μg/g
Zn900μg/g

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Chemical Name or Material 23 Element Oil Standard
Packaging Triple-leached LDPE bottle
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Analyte or Component Names Al,Ba,B
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Type Certified Reference Material
Element Al, Ba, B
Solvent or Matrix Base Oil 75
Industry Type Petrochemical Testing
Quantity 50 g

Product intended for laboratory use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.