missing translation for 'onlineSavingsMsg'
Learn More

Fisher Chemical™ Multi-element standard solution in Water for Ion Chromatography

Product Code. 31034068 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31034068 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 31034068 Supplier Fisher Chemical™ Supplier No. CM 8933
  • ONLINE EXCLUSIVE PRICE
    146.00 €
    131.56 € / 100mL
    Save 14.44 €
    9% Off
 Restricted Item
Estimated Shipment: 24-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Certified mixture of common anions used to calibrate IC instruments and ensure accurate, traceable results.

  • 6 components: 100 mg/L each of Fluorides (F-) ; Chlorides (Cl-) ; Bromides (Br-) ; Nitrates (NO3-) ; Phosphates (PO43-) ; Sulphates (SO42-).
  • Certified Reference Material (CRM) according to ISO 17025 and ISO 17034.
  • Calibration curves: Multi-element CRMs allow simultaneous calibration for several ions, saving time.
  • Quality control: Certified concentrations with documented uncertainty ensure traceability and reproducibility.
  • Regulatory compliance: Required in environmental, food, and pharmaceutical testing where ion chromatography is used.
  • Convenience: Ready-to-use solutions eliminate the need to prepare and standardize mixtures manually.
  • Each CRM comes with a certificate of analysis that includes uncertainty calculations.
  • This documentation is critical for audits, method validation, and proving the reliability of your data.

Specifications

Quantity 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.