missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Power Adapters for Digital Vortex Mixers

Product Code. 16338816 Shop All Thermo Scientific Products
Change view
Click to view available options
Description:
Multi-power adapter 220V
Plug Type:
EU, UK, CN Plug
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Plug Type unitSize
16338816 Multi-power adapter 220V EU, UK, CN Plug Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16338816 Supplier Thermo Scientific™ Supplier No. 88870124
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Power adapters for Digital Vortex Mixers

Power adapters for Digital Vortex Mixers, which offer accurate and efficient sample mixing

Service Plan

Standard Thermo warranty guideline, provide repair service and free spare part within warranty

Warranty

2 years

TRUSTED_SUSTAINABILITY

Specifications

Plug Type EU, UK, CN Plug
Material Plastic, cable, rubber, screw
Color Black
Description Multi-power adapter 220V
For Use With (Equipment) Digital Vortex Mixer
Includes Power adapter
Packaging Quantity 1 Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.