Learn More
Multi-State Hydrocarbon Standard, Hexane-Tetratetracontane Even Alkanes, High Level, SPEX CertiPrep™
Shop All SPEX Certiprep ProductsDescription
Includes
Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
| Analyte | CAS No. | Concentration |
|---|---|---|
| n-Decane | 124-18-5 | 1000μg/mL |
| n-Docosane | 629-97-0 | 1000μg/mL |
| n-Dodecane | 112-40-3 | 1000μg/mL |
| n-Dotetracontane | 7098-20-6 | 1000μg/mL |
| n-Dotriacontane | 544-85-4 | 1000μg/mL |
| n-Eicosane | 112-95-8 | 1000μg/mL |
| n-Hexacosane | 630-01-3 | 1000μg/mL |
| n-Hexadecane | 544-76-3 | 1000μg/mL |
| n-Hexane | 110-54-3 | 1000μg/mL |
| n-Hexatriacontane | 630-06-8 | 1000μg/mL |
| n-Octacosane | 630-02-4 | 1000μg/mL |
| n-Octadecane | 593-45-3 | 1000μg/mL |
| n-Octane | 111-65-9 | 1000μg/mL |
| n-Octatriacontane | 7194-85-6 | 1000μg/mL |
| n-Tetracontane | 4181-95-7 | 1000μg/mL |
| n-Tetracosane | 646-31-1 | 1000μg/mL |
| n-Tetradecane | 629-59-4 | 1000μg/mL |
| n-Tetratetracontane | 7098-22-8 | 1000μg/mL |
| n-Tetratriacontane | 14167-59-0 | 1000μg/mL |
| n-Triacontane | 638-68-6 | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Specifications
| Chemical Name or Material | Multi-State Hydrocarbon Standard (High Level) |
| Packaging | 2mL amber prescored glass ampule |
| Concentration or Composition Notes | Certificate of Analysis reports target analytes to a ±2% accuracy and correct for raw material purity. |
| For Use With (Equipment) | GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation |
| Type | Certified Reference Material |
| Solvent or Matrix | Cyclohexane:CS2 |
| Industry Type | Petrochemical Testing |
| Quantity | 1 mL |
Safety and Handling
- ALK MIX (C6-C44)
- Danger
- Aspiration toxicity Category 1
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 1
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- H361-Suspected of damaging fertility or the unborn child.
- H372-Causes damage to organs.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contain: cyclohexane, carbondisulphide, n-hexane
Product intended for laboratory use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.