missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Murashige and Skoog Media

Product Code. 11424865 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 x 1 L
1 x 10 L
Product Type:
Murashige and Skoog Medium
Content And Storage:
4°C
Unit Size:
10 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Product Type Content And Storage Unit Size
11424865 10 x 1 L Murashige and Skoog Medium 4°C 10 L
11434865 1 x 10 L Murashige and Skoog Medium 4°C 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11424865 Supplier MP Biomedicals™ Supplier No. 092610020
  • ONLINE EXCLUSIVE PRICE
    108.00 €
    101.85 € / 10 L
    Save 6.15 €
    5% Off
View all promotions
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

All Murashige and Skoog plant cell culture Media are manufactured from high quality chemicals and reagents.

  • Each formulation is prepared according to the original specification, except where noted
  • Finished lots of media are plant cell tested

Specifications

Form Powder
Product Type Murashige and Skoog Medium
Without Additives No Sucrose, No Agar, No IAA, No Kinetin
Quantity 10 x 1 L
Format Solution
pH 4.80 to 5.65
Content And Storage 4°C
compliance-icons compliance-icons
Product Identifier
  • Murashige and Skoog Medium, without Sucrose, Agar, IAA, and Kinetin
Signal Word
  • Danger
Hazard Category
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.