missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Muscarinic Acetylcholine Receptor M1/CHRM1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
271.00 € - 493.00 €
Specifications
| Antigen | Muscarinic Acetylcholine Receptor M1/CHRM1 |
|---|---|
| Dilution | Western Blot 1:1000 - 1:5000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18656182
|
Novus Biologicals
NBP2-94811-0.02ml |
0.02 mL |
271.00 €
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18665532
|
Novus Biologicals
NBP2-94811-0.1ml |
0.1 mL |
493.00 €
0.01mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Muscarinic Acetylcholine Receptor M1/CHRM1 Polyclonal antibody specifically detects Muscarinic Acetylcholine Receptor M1/CHRM1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
| Muscarinic Acetylcholine Receptor M1/CHRM1 | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cell Cycle and Replication, GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission, Transcription Factors and Regulators | |
| PBS (pH 7.3), 50% glycerol | |
| 1128 | |
| IgG | |
| Affinity purified |
| Western Blot 1:1000 - 1:5000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| cholinergic receptor, muscarinic 1, HM1, M1, M1R, MGC30125, muscarinic acetylcholine receptor M1 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Muscarinic Acetylcholine Receptor M1/CHRM1 (NP_000729.2). PPAVSPNITVLAPGKGPWQ | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title