Learn More
Invitrogen™ MVK Recombinant Rabbit Monoclonal Antibody (24GB115)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557476
 View more versions of this product
Product Code. 30276271
  • 564.00 € / 100µL
Description
MVK Recombinant Monoclonal Antibody for Western Blot, ICC/IF
Mevalonate kinase (MVK) is an early enzyme in isoprenoid and sterol synthesis. Mevalonate kinase catalyzes the ATP-dependent phosphorylation of mevalonic acid to form mevalonate 5-phosphate. Deficiency in MVK activity contributes to mevalonic aciduria and hyperimmunoglobulinemia D/periodic fever syndrome (HIDS).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MVK
Recombinant Monoclonal
Unconjugated
Mvk
2310010A05Rik; AI256848; AI414037; KIME; LH receptor mRNA-binding protein; LHR mRNA binding protein; Lrbp; Mevalonate kinase; mevalonate kinase 1; MK; Mvk; MVLK; POROK3
Rabbit
Affinity chromatography
RUO
4598, 81727
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB115
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P17256, Q03426
Mvk
A synthesized peptide derived from human MVK (aa 320-370).
100 μL
Primary
Human, Rat