missing translation for 'onlineSavingsMsg'
Learn More

MX69, MedChemExpress

Product Code. 16499756
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16499756 50 mg 50mg
16434486 100 mg 100mg
16415947 5 mg 5mg
16403218 10 mg 10mg
16400719 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16499756 Supplier Medchem Express Supplier No. HY100892

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress MX69 is an inhibitor of MDM2/XIAP, used for cancer treatment.

MX69 is an inhibitor of MDM2/XIAP, used for cancer treatment.

Specifications

Chemical Name or Material MX69
CAS 1005264-47-0
Color White
Purity Grade Notes Research
Molecular Formula C27H26N2O4S
Quantity 50 mg
Solubility Information DMSO : 100 mg/mL (210.72 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(O)C1=CC=C(C2C(CC=C3)C3C(C=C(S(=O)(NC4=CC(C)=C(C)C=C4)=O)C=C5)=C5N2)C=C1
Molecular Weight (g/mol) 474.57
Formula Weight 474.57
Percent Purity 99.65%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.