missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MYADM Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5144100
This item is not returnable.
View return policy
Description
MYADM Polyclonal Antibody for ICC/IF, IHC (P), ELISA
MYADM, belonging to the MAL family, is a 322-amino acid protein having 7 hydrophobic transmembrane domains, several potential consensus sites for phosphorylation and sequence homology with the T-cell differentiation protein MAL and plasmolipin. It shares 75.8% sequence identity with mouse in the transmembrane regions and is associated with differentiation of hematopoietic cells or acute promyelocytic leukemia cells. Its expression is up-regulated in NB4 cells induced by all-trans retinoic acid whereas down regulation inhibits colony formation of bone marrow cells. MYADM could serve as a new marker for hematopoietic differentiation and is a target for diagnostic, prognostic and/or therapeutic interventions. It is known to express widely in brain and pancreatic islet cells.
Specifications
| MYADM | |
| Polyclonal | |
| Unconjugated | |
| MYADM | |
| MYADM; myeloid associated differentiation marker; myeloid upregulated protein; myeloid up-regulated protein; myeloid-associated differentiation marker; Protein SB135; SB135; UNQ553/PRO1110 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 91663 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
| Q96S97 | |
| MYADM | |
| Recombinant Human Myeloid-associated differentiation marker protein (1-40AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction