missing translation for 'onlineSavingsMsg'
Learn More

Myclobutanil, MedChemExpress

Product Code. 16418856
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16418856 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16418856 Supplier Medchem Express Supplier No. HYB2148
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Myclobutanil is a conazole class fungicide widely used as an agrichemical.

Myclobutanil is a conazole class fungicide widely used as an agrichemical.

Specifications

Chemical Name or Material Myclobutanil
Physical Form Solid
CAS 88671-89-0
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C15H17ClN4
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : ≥ 100 mg/mL (346.28 mM)
SMILES N#CC(C1=CC=C(Cl)C=C1)(CCCC)CN2N=CN=C2
Molecular Weight (g/mol) 288.78
Formula Weight 288.78
Percent Purity 99.11%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.