missing translation for 'onlineSavingsMsg'
Learn More

Mycophenolate Mofetil, MedChemExpress

Product Code. 16498166
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
200 mg
1 g
5 g
Unit Size:
100mg
1g
200mg
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16498166 100 mg 100mg
16471396 200 mg 200mg
16462079 1 g 1g
16400818 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16498166 Supplier Medchem Express Supplier No. HYB0199
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Mycophenolate mofetil (RS 61443) is the morpholinoethylester prodrug of Mycophenolic acid. Mycophenolate mofetil inhibits de novo purine synthesis via the inhibition of inosine monophosphate dehydrogenase (IMPDH). Mycophenolate mofetil shows selective lymphocyte antiproliferative effects involve both T and B cells, preventing antibody formation.

Mycophenolate mofetil (RS 61443) is the morpholinoethylester prodrug of Mycophenolic acid. Mycophenolate mofetil inhibits de novo purine synthesis via the inhibition of inosine monophosphate dehydrogenase (IMPDH). Mycophenolate mofetil shows selective lymphocyte antiproliferative effects involve both T and B cells, preventing antibody formation.

Specifications

Chemical Name or Material Mycophenolate Mofetil
CAS 128794-94-5
Color White
Purity Grade Notes Research
Molecular Formula C23H31NO7
Quantity 100 mg
Synonym RS 61443 TM-MMF
Solubility Information DMSO : 100 mg/mL (230.69 mM; Need ultrasonic)
SMILES O=C(OCCN1CCOCC1)CC/C(C)=C/CC2=C(O)C3=C(COC3=O)C(C)=C2OC
Molecular Weight (g/mol) 433.49
Formula Weight 433.49
Percent Purity 99.48%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.